Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTTCGCCTAGACACGTTCCTGTC[A/G]CCTCTGACAGCTGCAGTTGCTCCCT
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 600909 MIM: 603294 | ||||||||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003906.4 | 6402 | Missense Mutation | GCG,GTG | A,V 1941 | NP_003897.2 | |
XM_005261203.4 | 6402 | Missense Mutation | GCG,GTG | A,V 1941 | XP_005261260.1 | |
XM_005261204.4 | 6402 | Missense Mutation | GCG,GTG | A,V 1941 | XP_005261261.1 | |
XM_005261205.3 | 6402 | Missense Mutation | GCG,GTG | A,V 1941 | XP_005261262.1 |
MCM3AP-AS1 - MCM3AP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |