Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTACGACAGGAGAGCTCCTCCAC[C/G]TTGCCCCTCGGGGGTTTCCAGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609196 MIM: 608865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105372779 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105372779 - uncharacterized LOC105372779 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRAP - melanocortin 2 receptor accessory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285394.1 | 461 | Silent Mutation | ACC,ACG | T,T 71 | NP_001272323.1 | |
NM_178817.3 | 461 | Silent Mutation | ACC,ACG | T,T 130 | NP_848932.1 | |
NM_206898.1 | 461 | Intron | NP_996781.1 | |||
XM_006724028.3 | 461 | Silent Mutation | ACC,ACG | T,T 130 | XP_006724091.1 | |
XM_017028407.1 | 461 | Intron | XP_016883896.1 |
URB1 - URB1 ribosome biogenesis 1 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014825.2 | 461 | UTR 3 | NP_055640.2 |