Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGATGAGAAGATTATGAATAGTTT[G/T]GAAAAGGGGAATAATTAATAGTGAT
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 609007 | |||||||||||||||||||||||||||||
Literature Links: |
LRRK2 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LRRK2 - leucine rich repeat kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198578.3 | Intron | NP_940980.3 | ||||
XM_005268629.3 | Intron | XP_005268686.1 | ||||
XM_011537877.2 | Intron | XP_011536179.1 | ||||
XM_011537881.2 | Intron | XP_011536183.1 | ||||
XM_011537882.2 | Intron | XP_011536184.1 | ||||
XM_017018786.1 | Intron | XP_016874275.1 | ||||
XM_017018787.1 | Intron | XP_016874276.1 | ||||
XM_017018788.1 | Intron | XP_016874277.1 | ||||
XM_017018789.1 | Intron | XP_016874278.1 |
Set Membership: |
HapMap |