Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTGGGACATCAGACCTTCGTGTC[C/T]CAGCCTGGCTATGTTGGGCTGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109135 | ||||||||||||||||||||
Literature Links: |
AXL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AXL - AXL receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278599.1 | 578 | Intron | NP_001265528.1 | |||
NM_001699.5 | 578 | Silent Mutation | TCC,TCT | S,S 128 | NP_001690.2 | |
NM_021913.4 | 578 | Silent Mutation | TCC,TCT | S,S 128 | NP_068713.2 |