Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTCCAGGAGGCGGAAGCAAGT[A/G]AGGCAGTAGAGACCTGGTGGAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607646 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCST2 - DC-STAMP domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144622.2 | 1427 | Silent Mutation | CTC,CTT | L,L 628 | NP_653223.2 | |
XM_011509188.2 | 1427 | Silent Mutation | CTC,CTT | L,L 408 | XP_011507490.1 | |
XM_011509189.2 | 1427 | Silent Mutation | CTC,CTT | L,L 296 | XP_011507491.1 |
ZBTB7B - zinc finger and BTB domain containing 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |