Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGTTGTAAGATCCCCAAAGACA[C/T]GGAAAATCCTGGACGAACAGATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607081 MIM: 185880 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAPBPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAPBPL - TAP binding protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018009.4 | 1731 | Intron | NP_060479.3 | |||
XM_005253700.3 | 1731 | Intron | XP_005253757.1 | |||
XM_017019545.1 | 1731 | Intron | XP_016875034.1 | |||
XM_017019546.1 | 1731 | Intron | XP_016875035.1 |
VAMP1 - vesicle associated membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297438.1 | 1731 | Intron | NP_001284367.1 | |||
NM_014231.4 | 1731 | UTR 3 | NP_055046.1 | |||
NM_016830.3 | 1731 | Intron | NP_058439.1 | |||
NM_199245.2 | 1731 | UTR 3 | NP_954740.1 |