Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAAATATTATTCAGAAGAAAA[C/T]TGACAGAGGGGAGGGAATGAGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF566 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF566 - zinc finger protein 566 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145343.1 | 4371 | UTR 3 | NP_001138815.1 | |||
NM_001145344.1 | 4371 | UTR 3 | NP_001138816.1 | |||
NM_001145345.1 | 4371 | UTR 3 | NP_001138817.1 | |||
NM_001300970.1 | 4371 | UTR 3 | NP_001287899.1 | |||
NM_032838.4 | 4371 | UTR 3 | NP_116227.1 | |||
XM_005259354.2 | 4371 | UTR 3 | XP_005259411.1 | |||
XM_005259356.2 | 4371 | UTR 3 | XP_005259413.1 | |||
XM_006723447.2 | 4371 | UTR 3 | XP_006723510.1 | |||
XM_011527428.1 | 4371 | UTR 3 | XP_011525730.1 | |||
XM_017027399.1 | 4371 | UTR 3 | XP_016882888.1 | |||
XM_017027400.1 | 4371 | UTR 3 | XP_016882889.1 |