Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTCATCGAACTGCGGCCATTTC[C/G]AACCCGCAATGGAAGAAAGGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 157147 MIM: 616013 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTTP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTTP - microsomal triglyceride transfer protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000253.3 | 10 | UTR 5 | NP_000244.2 | |||
NM_001300785.1 | 10 | UTR 5 | NP_001287714.1 |
TRMT10A - tRNA methyltransferase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134665.2 | 10 | Intron | NP_001128137.1 | |||
NM_001134666.2 | 10 | Intron | NP_001128138.1 | |||
NM_152292.4 | 10 | Intron | NP_689505.1 | |||
XM_005263352.3 | 10 | Intron | XP_005263409.1 | |||
XM_006714417.2 | 10 | Intron | XP_006714480.1 |
Set Membership: |
HapMap |