Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601199 | |||||||||||||||||||||||||||||||||||
Literature Links: |
CASR PubMed Links | |||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | ||||||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR - Not Available | ||||||||
AMR - Not Available |
CASR - calcium sensing receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000388.3 | 86 | Intron | NP_000379.2 | |||
NM_001178065.1 | 86 | UTR 5 | NP_001171536.1 | |||
XM_005247837.2 | 86 | Intron | XP_005247894.1 | |||
XM_006713789.3 | 86 | UTR 5 | XP_006713852.1 | |||
XM_017007324.1 | 86 | Intron | XP_016862813.1 | |||
XM_017007325.1 | 86 | Intron | XP_016862814.1 |
Set Membership: |
Validated |