Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605868 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP11A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP11A - ATPase phospholipid transporting 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015205.2 | Intron | NP_056020.2 | ||||
NM_032189.3 | Intron | NP_115565.3 | ||||
XM_005268299.4 | Intron | XP_005268356.1 | ||||
XM_005268300.4 | Intron | XP_005268357.1 | ||||
XM_005268301.4 | Intron | XP_005268358.1 | ||||
XM_005268303.4 | Intron | XP_005268360.1 | ||||
XM_005268305.4 | Intron | XP_005268362.1 | ||||
XM_005268306.4 | Intron | XP_005268363.1 | ||||
XM_011537480.2 | Intron | XP_011535782.1 | ||||
XM_017020490.1 | Intron | XP_016875979.1 | ||||
XM_017020491.1 | Intron | XP_016875980.1 | ||||
XM_017020492.1 | Intron | XP_016875981.1 |