Save up to 31% on select cell culture essentials

Bundle and save

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

SNP Genotyping
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › C___1799586_20
            See other IL17A GT Assays ›
            SNP ID:
            rs4711998
            Gene
            IL17A
            Gene Name
            interleukin 17A
            Set Membership:
            > HapMap
            Chromosome Location:
            Chr.6: 52185555 - 52185555 on Build GRCh38
            Polymorphism:
            A/G, Transition substitution
            Context Sequence [VIC/FAM]:

            GTATTCCTGAGAAGGAACTATTCTC[A/G]AGGACCTGAGTCCAAGTTCATCTTA

            Assay ID C___1799586_20
            Size
            Availability Made To Order
            Catalog # 4351379
            Price (USD) 442.00
            Your Price
            Online offer (USD):407.65
            407.65
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data...
            ×
            Back To Top

            Assay Details



            Species:

            Human

            dbSNP Submissions:

            NA

            Phenotype:

            MIM: 603149

            Literature Links:

            IL17A PubMed Links

            Allele Nomenclature:

            Minor Allele Frequency:

            1000Genome Applied Biosystems® HapMap
            Global
            A (0.48)
            (0.52)
            Caucasian - Not Available CEPH (CEU)
            A (0.22)
            (0.78)
            EAS
            G (0.23)
            (0.77)
            African American - Not Available YRI (Yoruba)
            G (0.37)
            (0.63)
            SAS
            A (0.45)
            (0.55)
            Chinese - Not Available JPT (Japanese)
            G (0.29)
            (0.71)
            AFR
            G (0.43)
            (0.57)
            Japanese - Not Available CHB (Han Chinese)
            G (0.23)
            (0.77)
            EUR
            A (0.26)
            (0.74)
            AMR
            A (0.25)
            (0.75)
            IL17A - interleukin 17A
            Transcript Accession SNP Location SNP Type Observed Codons Observed Amino Acid Protein ID
            NM_002190.2 Intron NP_002181.1

            Back To Top

            More Information


            Set Membership:

            HapMap

            Gene Ontology Categories:

            Function(s) Process(es)

            apoptotic process
            inflammatory response
            immune response
            cell surface receptor signaling pathway
            cell-cell signaling
            cell death
            positive regulation of necrotic cell death
            positive regulation of interleukin-23 production
            response to amino acid
            positive regulation of osteoclast differentiation
            positive regulation of transcription from RNA polymerase II promoter
            defense response to fungus
            cellular response to interleukin-1
            cellular response to glucocorticoid stimulus
            fibroblast activation
            positive regulation of cytokine production involved in inflammatory response
            positive regulation of interleukin-6 secretion
            cytokine activity
            cytokine receptor binding
            protein binding

            Back To Top

            Related Products

            • TaqMan® Genotyping Master Mix
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-6ff95d844f-5rtgr:80/100.66.79.192:80.
            git-commit: f44aa9153aef922f8fad9a05e87b8aee811da128
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.28.0-Offline