Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGGCTCCCATCCCAGGAGGTGG[A/C]GATGACATTACATTTCCGGGTTTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612175 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAB39L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAB39L - calcium binding protein 39 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079670.2 | Intron | NP_001073138.1 | ||||
NM_001287337.1 | Intron | NP_001274266.1 | ||||
NM_001287338.1 | Intron | NP_001274267.1 | ||||
NM_001287339.1 | Intron | NP_001274268.1 | ||||
NM_030925.3 | Intron | NP_112187.2 | ||||
XM_011535254.2 | Intron | XP_011533556.1 | ||||
XM_011535255.2 | Intron | XP_011533557.1 | ||||
XM_011535256.2 | Intron | XP_011533558.1 | ||||
XM_017020785.1 | Intron | XP_016876274.1 | ||||
XM_017020786.1 | Intron | XP_016876275.1 | ||||
XM_017020787.1 | Intron | XP_016876276.1 | ||||
XM_017020788.1 | Intron | XP_016876277.1 |