Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606136 | ||||||||||||||||||||
Literature Links: |
LINC00083 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00083 - long intergenic non-protein coding RNA 83 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017003086.1 | Intron | XP_016858575.1 |
RASAL2 - RAS protein activator like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004841.3 | Intron | NP_004832.1 | ||||
NM_170692.2 | Intron | NP_733793.2 | ||||
XM_005245622.3 | Intron | XP_005245679.1 | ||||
XM_011510166.2 | Intron | XP_011508468.1 | ||||
XM_011510167.2 | Intron | XP_011508469.1 | ||||
XM_017002849.1 | Intron | XP_016858338.1 | ||||
XM_017002850.1 | Intron | XP_016858339.1 | ||||
XM_017002851.1 | Intron | XP_016858340.1 | ||||
XM_017002852.1 | Intron | XP_016858341.1 | ||||
XM_017002853.1 | Intron | XP_016858342.1 | ||||
XM_017002854.1 | Intron | XP_016858343.1 | ||||
XM_017002855.1 | Intron | XP_016858344.1 |