Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAAACACGAACTACATTCTACA[A/G]TGCCATCCGTAACCAGAAATTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300552 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MID1 - midline 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000381.3 | Intron | NP_000372.1 | ||||
NM_001098624.2 | Intron | NP_001092094.1 | ||||
NM_001193277.1 | Intron | NP_001180206.1 | ||||
NM_001193278.1 | Intron | NP_001180207.1 | ||||
NM_001193279.1 | Intron | NP_001180208.1 | ||||
NM_001193280.1 | Intron | NP_001180209.1 | ||||
NM_001193281.1 | Intron | NP_001180210.1 | ||||
NM_033289.1 | Intron | NP_150631.1 | ||||
NM_033290.3 | Intron | NP_150632.1 | ||||
XM_005274536.1 | Intron | XP_005274593.1 | ||||
XM_005274537.1 | Intron | XP_005274594.1 | ||||
XM_006724492.3 | Intron | XP_006724555.1 | ||||
XM_011545525.2 | Intron | XP_011543827.1 | ||||
XM_011545526.2 | Intron | XP_011543828.1 | ||||
XM_011545527.2 | Intron | XP_011543829.1 | ||||
XM_017029534.1 | Intron | XP_016885023.1 | ||||
XM_017029535.1 | Intron | XP_016885024.1 | ||||
XM_017029536.1 | Intron | XP_016885025.1 | ||||
XM_017029537.1 | Intron | XP_016885026.1 | ||||
XM_017029538.1 | Intron | XP_016885027.1 | ||||
XM_017029539.1 | Intron | XP_016885028.1 | ||||
XM_017029540.1 | Intron | XP_016885029.1 | ||||
XM_017029541.1 | Intron | XP_016885030.1 | ||||
XM_017029542.1 | Intron | XP_016885031.1 |