Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACAGAGCTTTGTGTGTTCCCCT[A/G]TATTTAGAGATTCCACTGGGATACT
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
ENDOD1 PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ENDOD1 - endonuclease domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015036.2 | Intron | NP_055851.1 |