Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1024 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIAA1024 - KIAA1024 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015206.2 | 3993 | Intron | NP_056021.1 | |||
XM_011521392.1 | 3993 | Intron | XP_011519694.1 | |||
XM_011521393.2 | 3993 | Intron | XP_011519695.1 | |||
XM_011521395.2 | 3993 | Intron | XP_011519697.1 | |||
XM_011521396.2 | 3993 | Intron | XP_011519698.1 | |||
XM_017022027.1 | 3993 | Intron | XP_016877516.1 |
TMED3 - transmembrane p24 trafficking protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301203.1 | 3993 | Intron | NP_001288132.1 | |||
NM_007364.3 | 3993 | Intron | NP_031390.1 | |||
XM_017022040.1 | 3993 | UTR 3 | XP_016877529.1 | |||
XM_017022041.1 | 3993 | Intron | XP_016877530.1 |
Set Membership: |
HapMap |