Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607265 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLINT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLINT1 - clathrin interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195555.1 | 2303 | UTR 3 | NP_001182484.1 | |||
NM_001195556.1 | 2303 | UTR 3 | NP_001182485.1 | |||
NM_014666.3 | 2303 | UTR 3 | NP_055481.1 | |||
XM_011534701.2 | 2303 | UTR 3 | XP_011533003.1 | |||
XM_017010087.1 | 2303 | UTR 3 | XP_016865576.1 | |||
XM_017010088.1 | 2303 | Intron | XP_016865577.1 |