Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCCGCCGCAGCTCGGGGGCGC[C/G]GGGGGCGGCAGCAGCAGGCACGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600967 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F5 - E2F transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083588.1 | 169 | Silent Mutation | GCC,GCG | A,A 44 | NP_001077057.1 | |
NM_001083589.1 | 169 | Intron | NP_001077058.1 | |||
NM_001951.3 | 169 | Silent Mutation | GCC,GCG | A,A 44 | NP_001942.2 |
LOC102723322 - uncharacterized LOC102723322 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |