Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609729 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDZRN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PDZRN3 - PDZ domain containing ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303139.1 | 3327 | UTR 3 | NP_001290068.1 | |||
NM_001303140.1 | 3327 | UTR 3 | NP_001290069.1 | |||
NM_001303141.1 | 3327 | UTR 3 | NP_001290070.1 | |||
NM_001303142.1 | 3327 | UTR 3 | NP_001290071.1 | |||
NM_015009.2 | 3327 | UTR 3 | NP_055824.1 | |||
XM_017005942.1 | 3327 | UTR 3 | XP_016861431.1 |