Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM114A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM114A2 - family with sequence similarity 114 member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317993.1 | 2745 | UTR 3 | NP_001304922.1 | |||
NM_001317994.1 | 2745 | UTR 3 | NP_001304923.1 | |||
NM_001317995.1 | 2745 | UTR 3 | NP_001304924.1 | |||
NM_018691.3 | 2745 | UTR 3 | NP_061161.2 | |||
XM_005268360.4 | 2745 | UTR 3 | XP_005268417.1 | |||
XM_005268361.4 | 2745 | UTR 3 | XP_005268418.1 | |||
XM_011537540.2 | 2745 | UTR 3 | XP_011535842.1 | |||
XM_011537541.2 | 2745 | UTR 3 | XP_011535843.1 | |||
XM_011537542.2 | 2745 | UTR 3 | XP_011535844.1 | |||
XM_017008967.1 | 2745 | UTR 3 | XP_016864456.1 | |||
XM_017008968.1 | 2745 | UTR 3 | XP_016864457.1 |