Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDLRAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LDLRAD1 - low density lipoprotein receptor class A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010978.3 | 1670 | UTR 3 | NP_001010978.2 | |||
NM_001276392.1 | 1670 | UTR 3 | NP_001263321.1 | |||
NM_001276393.1 | 1670 | UTR 3 | NP_001263322.1 | |||
NM_001276394.1 | 1670 | UTR 3 | NP_001263323.1 | |||
NM_001276395.1 | 1670 | UTR 3 | NP_001263324.1 | |||
XM_011541446.1 | 1670 | Intron | XP_011539748.1 | |||
XM_017001264.1 | 1670 | UTR 3 | XP_016856753.1 |