Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGTGTTTAACAGCAGAGGTGCA[A/G]GGCGGGGGCTCACGTCCTGCTCCTC
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 604489 MIM: 606202 | ||||||||||||||||||||||||||
Literature Links: |
AMACR PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
AMACR - alpha-methylacyl-CoA racemase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167595.1 | 1076 | Missense Mutation | CCT,CTT | P,L 327 | NP_001161067.1 | |
NM_014324.5 | 1076 | Missense Mutation | CCT,CTT | P,L 327 | NP_055139.4 | |
NM_203382.2 | 1076 | UTR 3 | NP_976316.1 |
C1QTNF3-AMACR - C1QTNF3-AMACR readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC45A2 - solute carrier family 45 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |