Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
![]() |
Literature Links: |
COL26A1 PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL26A1 - collagen type XXVI alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278563.2 | Intron | NP_001265492.1 | ||||
NM_133457.4 | Intron | NP_597714.2 | ||||
XM_017011743.1 | Intron | XP_016867232.1 | ||||
XM_017011744.1 | Intron | XP_016867233.1 | ||||
XM_017011745.1 | Intron | XP_016867234.1 |
Set Membership: |
![]() |