Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616513 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LARP4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LARP4B - La ribonucleoprotein domain family member 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015155.2 | 7617 | UTR 3 | NP_055970.1 | |||
XM_005252432.3 | 7617 | Intron | XP_005252489.1 | |||
XM_005252435.3 | 7617 | Intron | XP_005252492.1 | |||
XM_011519435.2 | 7617 | Intron | XP_011517737.1 | |||
XM_017015987.1 | 7617 | Intron | XP_016871476.1 | |||
XM_017015988.1 | 7617 | Intron | XP_016871477.1 | |||
XM_017015989.1 | 7617 | Intron | XP_016871478.1 | |||
XM_017015990.1 | 7617 | Intron | XP_016871479.1 | |||
XM_017015991.1 | 7617 | Intron | XP_016871480.1 | |||
XM_017015992.1 | 7617 | Intron | XP_016871481.1 | |||
XM_017015993.1 | 7617 | Intron | XP_016871482.1 | |||
XM_017015994.1 | 7617 | Intron | XP_016871483.1 | |||
XM_017015995.1 | 7617 | Intron | XP_016871484.1 |
Set Membership: |
HapMap |