Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616773 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIGA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIGA1 - mitoguardin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270384.1 | Intron | NP_001257313.1 | ||||
NM_198549.3 | Intron | NP_940951.1 | ||||
XM_005270846.4 | Intron | XP_005270903.1 | ||||
XM_005270847.4 | Intron | XP_005270904.1 | ||||
XM_005270848.4 | Intron | XP_005270905.1 | ||||
XM_011541422.2 | Intron | XP_011539724.1 | ||||
XM_017001227.1 | Intron | XP_016856716.1 | ||||
XM_017001228.1 | Intron | XP_016856717.1 | ||||
XM_017001229.1 | Intron | XP_016856718.1 | ||||
XM_017001230.1 | Intron | XP_016856719.1 | ||||
XM_017001231.1 | Intron | XP_016856720.1 | ||||
XM_017001232.1 | Intron | XP_016856721.1 |