Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601925 MIM: 176790 | ||||||||||||||||||||
Literature Links: |
ARHGDIA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGDIA - Rho GDP dissociation inhibitor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185077.2 | 1150 | UTR 3 | NP_001172006.1 | |||
NM_001185078.2 | 1150 | UTR 3 | NP_001172007.1 | |||
NM_001301240.1 | 1150 | Intron | NP_001288169.1 | |||
NM_001301241.1 | 1150 | Intron | NP_001288170.1 | |||
NM_001301242.1 | 1150 | Missense Mutation | AAG,ACG | K,T 181 | NP_001288171.1 | |
NM_001301243.1 | 1150 | UTR 3 | NP_001288172.1 | |||
NM_004309.5 | 1150 | UTR 3 | NP_004300.1 | |||
XM_011523574.1 | 1150 | UTR 3 | XP_011521876.1 |
P4HB - prolyl 4-hydroxylase subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |