Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609075 MIM: 601053 | ||||||||||||||||||||
Literature Links: |
FBXW12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXW12 - F-box and WD repeat domain containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLXNB1 - plexin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130082.2 | 10483 | UTR 3 | NP_001123554.1 | |||
NM_002673.5 | 10483 | UTR 3 | NP_002664.2 | |||
XM_011533833.2 | 10483 | UTR 3 | XP_011532135.1 | |||
XM_011533834.1 | 10483 | UTR 3 | XP_011532136.1 | |||
XM_011533835.1 | 10483 | UTR 3 | XP_011532137.1 | |||
XM_011533836.1 | 10483 | UTR 3 | XP_011532138.1 | |||
XM_011533837.2 | 10483 | UTR 3 | XP_011532139.1 | |||
XM_017006630.1 | 10483 | UTR 3 | XP_016862119.1 | |||
XM_017006631.1 | 10483 | UTR 3 | XP_016862120.1 |