Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTCAGTTAAAGTTTTGAACAC[A/G]TAATCCTTTTCAGTCAACTTACTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
YTHDF3 - YTH N6-methyladenosine RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277813.1 | Intron | NP_001264742.1 | ||||
NM_001277814.1 | Intron | NP_001264743.1 | ||||
NM_001277815.1 | Intron | NP_001264744.1 | ||||
NM_001277816.1 | Intron | NP_001264745.1 | ||||
NM_001277817.1 | Intron | NP_001264746.1 | ||||
NM_001277818.1 | Intron | NP_001264747.1 | ||||
NM_152758.5 | Intron | NP_689971.4 | ||||
XM_011517509.2 | Intron | XP_011515811.1 | ||||
XM_011517510.2 | Intron | XP_011515812.1 |
YTHDF3-AS1 - YTHDF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |