Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603772 | ||||||||||||||||||||
Literature Links: |
DYNC1I1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DYNC1I1 - dynein cytoplasmic 1 intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135556.1 | 642 | Intron | NP_001129028.1 | |||
NM_001135557.1 | 642 | Intron | NP_001129029.1 | |||
NM_001278421.1 | 642 | Intron | NP_001265350.1 | |||
NM_001278422.1 | 642 | Intron | NP_001265351.1 | |||
NM_004411.4 | 642 | Intron | NP_004402.1 | |||
XM_011515861.1 | 642 | Intron | XP_011514163.1 | |||
XM_011515862.1 | 642 | Intron | XP_011514164.1 | |||
XM_011515863.2 | 642 | Intron | XP_011514165.1 | |||
XM_017011804.1 | 642 | UTR 5 | XP_016867293.1 | |||
XM_017011805.1 | 642 | Intron | XP_016867294.1 | |||
XM_017011806.1 | 642 | Intron | XP_016867295.1 | |||
XM_017011807.1 | 642 | Intron | XP_016867296.1 |