Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616434 MIM: 607895 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IST1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IST1 - IST1, ESCRT-III associated factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD1L3 - polycystin 1 like 3, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181536.1 | Intron | NP_853514.1 | ||||
XM_017023201.1 | Intron | XP_016878690.1 | ||||
XM_017023202.1 | Intron | XP_016878691.1 | ||||
XM_017023203.1 | Intron | XP_016878692.1 | ||||
XM_017023204.1 | Intron | XP_016878693.1 |
Set Membership: |
HapMap |