Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608918 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATPAF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATPAF2 - ATP synthase mitochondrial F1 complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GID4 - GID complex subunit 4 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024052.4 | 508 | Silent Mutation | GGA,GGG | G,G 151 | NP_076957.3 | |
XM_005256799.4 | 508 | Silent Mutation | GGA,GGG | G,G 151 | XP_005256856.1 | |
XM_005256800.1 | 508 | Silent Mutation | GGA,GGG | G,G 151 | XP_005256857.1 | |
XM_011524008.1 | 508 | Silent Mutation | GGA,GGG | G,G 151 | XP_011522310.1 |