Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 132350 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STX2 - syntaxin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001980.3 | 3017 | UTR 3 | NP_001971.2 | |||
NM_194356.2 | 3017 | UTR 3 | NP_919337.1 | |||
XM_005253557.1 | 3017 | UTR 3 | XP_005253614.1 | |||
XM_017018978.1 | 3017 | UTR 3 | XP_016874467.1 | |||
XM_017018979.1 | 3017 | UTR 3 | XP_016874468.1 | |||
XM_017018980.1 | 3017 | UTR 3 | XP_016874469.1 | |||
XM_017018981.1 | 3017 | Intron | XP_016874470.1 | |||
XM_017018982.1 | 3017 | UTR 3 | XP_016874471.1 | |||
XM_017018983.1 | 3017 | UTR 3 | XP_016874472.1 | |||
XM_017018984.1 | 3017 | UTR 3 | XP_016874473.1 | |||
XM_017018985.1 | 3017 | UTR 3 | XP_016874474.1 | |||
XM_017018986.1 | 3017 | Intron | XP_016874475.1 | |||
XM_017018987.1 | 3017 | Intron | XP_016874476.1 | |||
XM_017018988.1 | 3017 | Intron | XP_016874477.1 | |||
XM_017018989.1 | 3017 | Intron | XP_016874478.1 |
Set Membership: |
HapMap Validated |