Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCAACACTGCCCGAGGCTGAC[T/C]GAGAGCGAGGTGCCATCATGGGCAT
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
![]() ![]() |
Literature Links: |
ACTN3 PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACTN3 - actinin alpha 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104.3 | 1974 | Nonsense Mutation | CGA,TGA | R,* 577 | NP_001095.2 | |
NM_001258371.2 | 1974 | Nonsense Mutation | CGA,TGA | R,* 620 | NP_001245300.2 |
CTSF - cathepsin F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |