Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACAGTCTGAATGACAGTCAAGC[T/A]GAAGGAGTTTTTAGGGTTGTGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601655 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EYA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EYA3 - EYA transcriptional coactivator and phosphatase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282560.1 | 2802 | Intron | NP_001269489.1 | |||
NM_001282561.1 | 2802 | UTR 3 | NP_001269490.1 | |||
NM_001282562.1 | 2802 | UTR 3 | NP_001269491.1 | |||
NM_001990.3 | 2802 | UTR 3 | NP_001981.2 | |||
XM_006710449.3 | 2802 | UTR 3 | XP_006710512.1 | |||
XM_011540999.2 | 2802 | Intron | XP_011539301.1 | |||
XM_011541001.2 | 2802 | Intron | XP_011539303.1 | |||
XM_011541003.2 | 2802 | Intron | XP_011539305.1 | |||
XM_011541004.2 | 2802 | UTR 3 | XP_011539306.1 | |||
XM_017000652.1 | 2802 | Intron | XP_016856141.1 | |||
XM_017000653.1 | 2802 | Intron | XP_016856142.1 | |||
XM_017000654.1 | 2802 | Intron | XP_016856143.1 | |||
XM_017000655.1 | 2802 | Intron | XP_016856144.1 | |||
XM_017000656.1 | 2802 | Intron | XP_016856145.1 | |||
XM_017000657.1 | 2802 | Intron | XP_016856146.1 | |||
XM_017000658.1 | 2802 | Intron | XP_016856147.1 | |||
XM_017000659.1 | 2802 | Intron | XP_016856148.1 |
XKR8 - XK related 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |