Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 72 |
Chromosome Location: | Chr. 1 - 198859348 - 198859348 [-] on 198859348 Build GRCh38 |
Species: | Human |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-181a-5p | MIMAT0000256 |
Human | hsa-miR-181a-3p | MIMAT0000270 |