Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 81 |
Chromosome Location: | Chr. 2 - 38853038 - 38853038 [+] on 38853038 Build GRCm38 |
Species: | Mouse |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-181a-5p | MIMAT0000210 |
Mouse | mmu-miR-181a-2-3p | MIMAT0005443 |