Invitrogen antibodies—maximize lab budgets with advanced validation & coverage

Shop antibodies

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Gene Expression
  • Microbe & ABR Detection
  • Copy Number Variation
  • Mutation Detection
  • miRNA
  • SNP Genotyping
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central
            This product has been added to your favorites list. Go to My Favorites

            System Message

            OKCancel
            LOADING ...
            • Home
            • › Search Tool
            • › Search Results
            • › Mm03306644_pri
            Assay Name:
            mmu-mir-301a
            Stem-loop ID:
            mmu-mir-301a
            Mapped to miRBase Version:
            v22.1
            Chromosome Location:
            Chr.11: 87113004 - 87113089 [+] on Build GRCm38
            Stem-loop Location:
            Chr.11: 87113004 - 87113089 [+] on Build GRCm38
            Mature miRNA ID:
            mmu-miR-301a-5p mmu-miR-301a-3p
            Species:
            Mouse
            Product Type:
            TaqMan™ Pri-miRNA Assay
            Assay ID Mm03306644_pri
            Size
            Availability Made To Order
            Catalog # 4427012
            Price
            Your Price
            Online offer:
            Check your price ›
            • Genomic Map
            • Assay Details
            • More Information

            Genomic Map

            LOADING... Fetching data..
            ×
            Back To Top

            Assay Details

            Amplicon Length:

            71

            Chromosome Location:

            Chr. 11 - 87112808 - 87112808 [-] on 87112808 Build GRCm38

            Species:

            Mouse

            miRBase Details:



            Species miRBase ID miRBase Accession Number
            Mouse mmu-miR-301a-5p MIMAT0017008
            Mouse mmu-miR-301a-3p MIMAT0000379

            Stem-loop Details



            Mouse

            Stem-loop ID mmu-mir-301a
            Stem-loop Accession # MI0000401
            Stem-loop Sequence
            CCUGCUAACGGCUGCUCUGACUUUAUUGCACUACUGUACUUUACAGCGAGCAGUGCAAUAGUAUUGUCAAAGCAUCCGCGAGCAGG
            Chromosome Location Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38
            Mature MicroRNA miRBase Accession #: MIMAT0017008
            miRBase ID: mmu-miR-301a-5p
            Mature miRNA Sequence: GCUCUGACUUUAUUGCACUACU
            Chromosome Location: Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38

            miRBase Accession #: MIMAT0000379
            miRBase ID: mmu-miR-301a-3p
            Mature miRNA Sequence: CAGUGCAAUAGUAUUGUCAAAGC
            Chromosome Location: Chr. 11 - 87113004 - 87113089 [+] on Build GRCm38

            Gene Family ID MIPF0000034, mir-130
            Distance to Amplicon 166 bases

            Back To Top

            More Information




            Related Products

            Ambion® Anti-miR™ miRNA Inhibitor : AM14414
            Ambion® Pre-miR™ miRNA Precursor : PM14414
            TaqMan™ MicroRNA Assay : 006346_mat
            mirVana® miRNA inhibitor : MH14414
            mirVana® miRNA mimic : MC14414
            TaqMan™ Advanced miRNA Assay : 478796_mir
            TaqMan™ Advanced miRNA Assay : mmu483007_mir
            Ambion® Anti-miR™ miRNA Inhibitor : AM10978
            Ambion® Pre-miR™ miRNA Precursor : PM10978
            TaqMan™ MicroRNA Assay : 000528
            mirVana® miRNA inhibitor : MH10978
            mirVana® miRNA mimic : MC10978
            TaqMan™ Advanced miRNA Assay : 477815_mir

            Back To Top

            Related Products

            • High Capacity RNA-to-cDNA Kit
            • SuperScript® IV VILO™ Master Mix
            • TaqMan® Universal PCR Master Mix
            • TaqMan® Fast Advanced Master Mix
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-nfgkx:80/100.66.77.49:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0