Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 89 |
Chromosome Location: | Chr. 3 - 23150800 - 23150800 [+] on 23150800 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-181a-5p | MIMAT0000858 |
Rat | rno-miR-181a-2-3p | MIMAT0017138 |