Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Amplicon Length: | 60 |
Chromosome Location: | Chr. 13 - 54952595 - 54952595 [-] on 54952595 Build Rnor_6.0 |
Species: | Rat |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-181a-5p | MIMAT0000858 |
Rat | rno-miR-181a-1-3p | MIMAT0000884 |
Rat | rno-miR-3570 | MIMAT0017850 |