Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-20a |
---|---|
miRBase Accession Number: | MI0000076 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UAAAGUGCUUAUAGUGCAGGUAG |
Species: | Human, Mouse, Rat, Alligator mississippiensis, Astatotilapia burtoni, Bovine, Callithrix jacchus, Chicken, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Dog, Fugu rubripes, Gadus morhua, Goat, Guinea pig, Horse, Metriaclima zebra, Microcebus murinus, Monodelphis domestica, Neolamprologus brichardi, Nomascus leucogenys, Ophiophagus hannah, Oreochromis niloticus, Pteropus alecto, Pundamilia nyererei, Python bivittatus, Rabbit, Salmo salar, Taeniopygia guttata, Tetraodon nigroviridis, Xenopus laevis, Xenopus tropicalis, Zebrafish |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UAAAGUGCUUAUAGUGCAGGUAG |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-20a-5p | MIMAT0000075 | |
Mouse | mmu-miR-20a-5p | MIMAT0000529 | |
Rat | rno-miR-20a-5p | MIMAT0000602 | |
Alligator mississippiensis | ami-miR-20a-5p | MIMAT0038054 | |
Astatotilapia burtoni | abu-miR-20a | MIMAT0042128 | |
Bovine | bta-miR-20a | MIMAT0003527 | |
View More Rows | |||
Callithrix jacchus | cja-miR-20a | MIMAT0039421 | |
Chicken | gga-miR-20a-5p | MIMAT0001111 | |
Cricetulus griseus | cgr-miR-20a | MIMAT0023847 | |
Cyprinus carpio | ccr-miR-20a-5p | MIMAT0026265 | |
Dasypus novemcinctus | dno-miR-20a-5p | MIMAT0047520 | |
Dog | cfa-miR-20a | MIMAT0006651 | |
Fugu rubripes | fru-miR-20 | MIMAT0003083 | |
Gadus morhua | gmo-miR-20a-5p | MIMAT0044245 | |
Goat | chi-miR-20a-5p | MIMAT0036054 | |
Guinea pig | cpo-miR-20a-5p | MIMAT0046868 | |
Horse | eca-miR-20a | MIMAT0013088 | |
Metriaclima zebra | mze-miR-20a | MIMAT0042358 | |
Microcebus murinus | mmr-miR-20 | MIMAT0049196 | |
Monodelphis domestica | mdo-miR-20a-5p | MIMAT0004169 | |
Neolamprologus brichardi | nbr-miR-20a | MIMAT0042539 | |
Nomascus leucogenys | nle-miR-20 | MIMAT0049453 | |
Ophiophagus hannah | oha-miR-20a-5p | MIMAT0036822 | |
Oreochromis niloticus | oni-miR-20 | MIMAT0042719 | |
Pteropus alecto | pal-miR-20a-5p | MIMAT0040138 | |
Pundamilia nyererei | pny-miR-20a | MIMAT0042908 | |
Python bivittatus | pbv-miR-20a-5p | MIMAT0038846 | |
Rabbit | ocu-miR-20a-5p | MIMAT0048120 | |
Salmo salar | ssa-miR-20a-5p | MIMAT0032502 | |
Taeniopygia guttata | tgu-miR-20a-5p | MIMAT0014553 | |
Tetraodon nigroviridis | tni-miR-20 | MIMAT0003084 | |
Xenopus laevis | xla-miR-20a-5p | MIMAT0046528 | |
Xenopus tropicalis | xtr-miR-20a-5p | MIMAT0003669 | |
Zebrafish | dre-miR-20a-5p | MIMAT0001786 |