Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-486 |
---|---|
miRBase Accession Number: | MI0002470 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.8: 41660441 - 41660508 [-] on Build GRCh38 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGAG |
Species: | Human, Mouse, Rat, Bovine, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rhesus monkey |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000220, mir-486 |
Mature miRNA Sequence: | UCCUGUACUGAGCUGCCCCGAG |
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-486-5p | MIMAT0002177 | |
Mouse | mmu-miR-486a-5p | MIMAT0003130 | |
Mouse | mmu-miR-486b-5p | MIMAT0014943 | |
Rat | rno-miR-486 | MIMAT0037265 | |
Bovine | bta-miR-486 | MIMAT0009329 | |
Cricetulus griseus | cgr-miR-486-5p | MIMAT0023967 | |
View More Rows | |||
Dasypus novemcinctus | dno-miR-486-5p | MIMAT0047912 | |
Gorilla gorilla | ggo-miR-486 | MIMAT0024149 | |
Guinea pig | cpo-miR-486-5p | MIMAT0047277 | |
Horse | eca-miR-486-5p | MIMAT0013186 | |
Pan troglodytes | ptr-miR-486 | MIMAT0008160 | |
Pig | ssc-miR-486 | MIMAT0013886 | |
Pongo pygmaeus | ppy-miR-486-5p | MIMAT0015904 | |
Pteropus alecto | pal-miR-486a-5p | MIMAT0039846 | |
Rhesus monkey | mml-miR-486-5p | MIMAT0006344 |
Stem-loop ID | hsa-mir-486-1 |
Stem-loop Accession # | MI0002470 |
Stem-loop Sequence | GCAUCCUGUACUGAGCUGCCCCGAGGCCCUUCAUGCUGCCCAGCUCGGGGCAGCUCAGUACAGGAUAC |
Chromosome Location | Chr. 8 - 41660441 - 41660508 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0002177
miRBase ID: hsa-miR-486-5p Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG Chromosome Location: Chr. 8 - 41660444 - 41660507 [+] on Build GRCh38 |
Stem-loop ID | hsa-mir-486-2 |
Stem-loop Accession # | MI0023622 |
Stem-loop Sequence | UCCUGUACUGAGCUGCCCCGAGCUGGGCAGCAUGAAGGGCCUCGGGGCAGCUCAGUACAGGAUG |
Chromosome Location | Chr. 8 - 41660444 - 41660507 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0002177
miRBase ID: hsa-miR-486-5p Mature miRNA Sequence: UCCUGUACUGAGCUGCCCCGAG Chromosome Location: Chr. 8 - 41660444 - 41660507 [+] on Build GRCh38 |