Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-92a-1* |
---|---|
miRBase Accession Number: | MI0000093 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGGUUGGGAUCGGUUGCAAUGCU |
Species: | Human, Guinea pig, Ophiophagus hannah, Pteropus alecto |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000013, mir-25 |
Mature miRNA Sequence: | AGGUUGGGAUCGGUUGCAAUGCU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-92a-1-5p | MIMAT0004507 |
Guinea pig | cpo-miR-92a-5p | MIMAT0046928 |
Ophiophagus hannah | oha-miR-92a-5p | MIMAT0036958 |
Pteropus alecto | pal-miR-92a-2-5p | MIMAT0039838 |