Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-17* |
---|---|
miRBase Accession Number: | MI0000687 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | ACUGCAGUGAGGGCACUUGUAG |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | ACUGCAGUGAGGGCACUUGUAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-17-3p | MIMAT0000650 |