Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-670 |
---|---|
miRBase Accession Number: | MI0003933 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.11: 43559656 - 43559753 [+] on Build GRCh38 |
Mature miRNA Sequence: | GUCCCUGAGUGUAUGUGGUG |
Species: | Human |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000734, mir-670 |
Mature miRNA Sequence: | GUCCCUGAGUGUAUGUGGUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-670-5p | MIMAT0010357 |
Stem-loop ID | hsa-mir-670 |
Stem-loop Accession # | MI0003933 |
Stem-loop Sequence | GUUUAGGGGUGGACCUGAUGUCCCUGAGUGUAUGUGGUGAACCUGAAUUUGCCUUGGGUUUCCUCAUAUUCAUUCAGGAGUGUCAGUUGCCCCUUCAC |
Chromosome Location | Chr. 11 - 43559656 - 43559753 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0010357
miRBase ID: hsa-miR-670-5p Mature miRNA Sequence: GUCCCUGAGUGUAUGUGGUG Chromosome Location: Chr. 11 - 43559656 - 43559753 [+] on Build GRCh38 |