Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-3115 |
---|---|
miRBase Accession Number: | MI0014127 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.1: 23044305 - 23044372 [+] on Build GRCh38 |
Mature miRNA Sequence: | AUAUGGGUUUACUAGUUGGU |
Species: | Human |
Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | AUAUGGGUUUACUAGUUGGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3115 | MIMAT0014977 |
Stem-loop ID | hsa-mir-3115 |
Stem-loop Accession # | MI0014127 |
Stem-loop Sequence | UCUGAAUAUGGGUUUACUAGUUGGUGGUGAAUUCAUGAGUCGCCAACUAUUAGGCCUUUAUGUCCAGA |
Chromosome Location | Chr. 1 - 23044305 - 23044372 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0014977
miRBase ID: hsa-miR-3115 Mature miRNA Sequence: AUAUGGGUUUACUAGUUGGU Chromosome Location: Chr. 1 - 23044305 - 23044372 [+] on Build GRCh38 |