Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-19b-2* |
---|---|
miRBase Accession Number: | MI0000546 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AGUUUUGCAGAUUUGCAGUUCAGC |
Species: | Mouse |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000011, mir-19 |
Mature miRNA Sequence: | AGUUUUGCAGAUUUGCAGUUCAGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-19b-2-5p | MIMAT0017010 |