Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-23a-3p |
---|---|
Stem-loop Accession Number: | MI0000079 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.19: 13836587 - 13836659 [-] on Build GRCh38 |
Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUUCC |
Species: | Human, Mouse, Rat, Anolis carolinensis, Ateles geoffroyi, Chrysemys picta, Cricetulus griseus, Cyprinus carpio, Goat, Gorilla gorilla, Horse, Ictalurus punctatus, Lemur catta, Macaca nemestrina, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pundamilia nyererei, Python bivittatus, Rhesus monkey, Saguinus labiatus, Salmo salar, Xenopus laevis, Xenopus tropicalis |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | AUCACAUUGCCAGGGAUUUCC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-23a-3p | MIMAT0000078 |
Mouse | mmu-miR-23a-3p | MIMAT0000532 |
Rat | rno-miR-23a-3p | MIMAT0000792 |
Anolis carolinensis | aca-miR-23a-3p | MIMAT0021896 |
Ateles geoffroyi | age-miR-23a | MIMAT0002741 |
Chrysemys picta | cpi-miR-23a-3p | MIMAT0037655 |
View More Rows | ||
Cricetulus griseus | cgr-miR-23a-3p | MIMAT0023863 |
Cyprinus carpio | ccr-miR-23a | MIMAT0026277 |
Goat | chi-miR-23a | MIMAT0036087 |
Gorilla gorilla | ggo-miR-23a | MIMAT0002738 |
Horse | eca-miR-23a | MIMAT0012986 |
Ictalurus punctatus | ipu-miR-23a | MIMAT0029481 |
Lemur catta | lca-miR-23a | MIMAT0002745 |
Macaca nemestrina | mne-miR-23a | MIMAT0002756 |
Pan paniscus | ppa-miR-23a | MIMAT0002743 |
Pan troglodytes | ptr-miR-23a | MIMAT0002749 |
Papio hamadryas | pha-miR-23a | MIMAT0049533 |
Pig | ssc-miR-23a | MIMAT0002133 |
Pongo pygmaeus | ppy-miR-23a | MIMAT0002747 |
Pundamilia nyererei | pny-miR-23a | MIMAT0042955 |
Python bivittatus | pbv-miR-23a-3p | MIMAT0038853 |
Rhesus monkey | mml-miR-23a-3p | MIMAT0002752 |
Saguinus labiatus | sla-miR-23a | MIMAT0002754 |
Salmo salar | ssa-miR-23a-3p | MIMAT0032549 |
Xenopus laevis | xla-miR-23a | MIMAT0011145 |
Xenopus tropicalis | xtr-miR-23a | MIMAT0003568 |
Stem-loop ID | hsa-mir-23a |
Stem-loop Accession # | MI0000079 |
Stem-loop Sequence | GGCCGGCUGGGGUUCCUGGGGAUGGGAUUUGCUUCCUGUCACAAAUCACAUUGCCAGGGAUUUCCAACCGACC |
Chromosome Location | Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000078
miRBase ID: hsa-miR-23a-3p Mature miRNA Sequence: AUCACAUUGCCAGGGAUUUCC Chromosome Location: Chr. 19 - 13836587 - 13836659 [-] on Build GRCh38 |