Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-184 |
---|---|
miRBase Accession Number: | MI0000481 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.15: 79209788 - 79209871 [+] on Build GRCh38 |
Mature miRNA Sequence: | UGGACGGAGAACUGAUAAGGGU |
Species: | Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Bovine, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Guinea pig, Horse, Macaca nemestrina, Monodelphis domestica, Otolemur garnettii, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Taeniopygia guttata, Tupaia chinensis |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000059, mir-184 |
Mature miRNA Sequence: | UGGACGGAGAACUGAUAAGGGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-184 | MIMAT0000454 |
Mouse | mmu-miR-184-3p | MIMAT0000213 |
Rat | rno-miR-184 | MIMAT0000861 |
Alligator mississippiensis | ami-miR-184-3p | MIMAT0038199 |
Anolis carolinensis | aca-miR-184-3p | MIMAT0021814 |
Bovine | bta-miR-184 | MIMAT0009246 |
View More Rows | ||
Callithrix jacchus | cja-miR-184 | MIMAT0039492 |
Chicken | gga-miR-184-3p | MIMAT0001158 |
Chrysemys picta | cpi-miR-184-3p | MIMAT0037801 |
Columba livia | cli-miR-184-3p | MIMAT0038561 |
Cricetulus griseus | cgr-miR-184 | MIMAT0023809 |
Dasypus novemcinctus | dno-miR-184-3p | MIMAT0047704 |
Dog | cfa-miR-184 | MIMAT0009842 |
Goat | chi-miR-184 | MIMAT0036009 |
Guinea pig | cpo-miR-184-3p | MIMAT0047053 |
Horse | eca-miR-184 | MIMAT0012894 |
Macaca nemestrina | mne-miR-184 | MIMAT0002303 |
Monodelphis domestica | mdo-miR-184-3p | MIMAT0004119 |
Otolemur garnettii | oga-miR-184 | MIMAT0049718 |
Pan troglodytes | ptr-miR-184 | MIMAT0002301 |
Pig | ssc-miR-184 | MIMAT0002127 |
Pongo pygmaeus | ppy-miR-184 | MIMAT0002302 |
Pteropus alecto | pal-miR-184-3p | MIMAT0040165 |
Rabbit | ocu-miR-184-3p | MIMAT0048290 |
Rhesus monkey | mml-miR-184 | MIMAT0006218 |
Taeniopygia guttata | tgu-miR-184 | MIMAT0014561 |
Tupaia chinensis | tch-miR-184 | MIMAT0036493 |
Stem-loop ID | hsa-mir-184 |
Stem-loop Accession # | MI0000481 |
Stem-loop Sequence | CCAGUCACGUCCCCUUAUCACUUUUCCAGCCCAGCUUUGUGACUGUAAGUGUUGGACGGAGAACUGAUAAGGGUAGGUGAUUGA |
Chromosome Location | Chr. 15 - 79209788 - 79209871 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0000454
miRBase ID: hsa-miR-184 Mature miRNA Sequence: UGGACGGAGAACUGAUAAGGGU Chromosome Location: Chr. 15 - 79209788 - 79209871 [+] on Build GRCh38 |