Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | mmu-miR-363-5p |
---|---|
miRBase Accession Number: | MI0000765 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAGGUGGAACACGAUGCAAUUU |
Species: | Mouse |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000138, mir-363 |
Mature miRNA Sequence: | CAGGUGGAACACGAUGCAAUUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-363-5p | MIMAT0017076 |