Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay Name: | hsa-miR-4531 |
---|---|
miRBase Accession Number: | MI0016898 |
miRBase Version: | v22.1 |
Chromosome Location: | Chr.19: 44653686 - 44653732 [-] on Build GRCh38 |
Mature miRNA Sequence: | AUGGAGAAGGCUUCUGA |
Species: | Human |
Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AUGGAGAAGGCUUCUGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4531 | MIMAT0019070 |
Stem-loop ID | hsa-mir-4531 |
Stem-loop Accession # | MI0016898 |
Stem-loop Sequence | GCCUAGGAGUCCUUGGUCAGUGGGGACAUGGAGAAGGCUUCUGAGGA |
Chromosome Location | Chr. 19 - 44653686 - 44653732 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019070
miRBase ID: hsa-miR-4531 Mature miRNA Sequence: AUGGAGAAGGCUUCUGA Chromosome Location: Chr. 19 - 44653686 - 44653732 [-] on Build GRCh38 |